Please login to your online account to display your discounted pricing

Potassium Bromide, 40g + 100mL Water, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Bromide
Name Note 40g + 100mL Water
CAS 7732-18-5
CAS 7758-02-3
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

R58800001A

 
ricca chemical company
RIR58800001A
Poly natural 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R58800004A

 
ricca chemical company
RIR58800004A
Poly natural 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 1L
Chemical Name or Material Potassium Bromide
CAS 7732-18-5
Odor Odorless
Packaging Poly natural
Solvent or Matrix Aqueous Solution
Name Note 40g + 100mL Water
CAS 7758-02-3