missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium bromate, Acculute Standard Volumetric Solution, Final Concentration 0.1N

Manufacturer:  Alfa Aesar™ 88592LQ

 View more versions of this product

Catalog No. AA88592LQ



Specifications

Specifications

Potassium bromate Acculute Standard Volumetric Solution
7758-01-2,7732-18-5
MFCD00011359
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
[O-]Br(=O)=O.[K+]
167.00
CHEBI:38211
Liquid
1 unit
Final Concentration 0.1N
BrKO3
UN3139
OCATYIAKPYKMPG-UHFFFAOYSA-M
potassium;bromate
23673461
167
Colorless

Chemical Identifiers

7758-01-2
167.00
OCATYIAKPYKMPG-UHFFFAOYSA-M
23673461
potassium;bromate
BrKO3
MFCD00011359
potassium bromate, bromic acid, potassium salt, bromate, potassium, potassium bromate dot, unii-04mb35w6za, ccris 529, eec no. e924, bromic acid, potassium salt 1:1, potassiumbromate, kaliumbromat
CHEBI:38211
[O-]Br(=O)=O.[K+]
Safety and Handling

Safety and Handling

GHS H Statement
H271-H350
May cause fire or explosion
strong oxidizer.
May cause cancer.

GHS P Statement
P221-P283-P210-P306+P360-P405-P501a
Take any precaution to avoid mixing with combustibles.
Wear fire/flame resistant/retardant clothing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON CLOTHING: rinse immediately contaminated clothing and skin with plenty of water before removing clothes.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: OXIDIZING LIQUID, N.O.S.

TSCA : TSCA

SDS Documents