Learn More
Potassium bromate, 99.5%, for analysis, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 268390010
Description
C-X bond formation reagent, Oxidation reagentSpecifications
Potassium bromate | |
7758-01-2 | |
BrKO3 | |
MFCD00011359 | |
15, 7738 | |
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol,soluble in boiling water | |
[O-]Br(=O)=O.[K+] | |
167.01 | |
CHEBI:38211 | |
Crystalline Powder | |
Analysis | |
3.2700g/mL | |
0.005% max. | |
5ppm max. | |
Plastic bottle | |
Solubility in water: 70g/L (20°C). Other solubilities: practically insoluble in alcohol, soluble in boiling water | |
50ppm max. | |
White | |
5 to 9 (5% soln. at 20°C) |
p.a. | |
99.5% min. (Iodometry) | |
KBrO3 | |
11,431 | |
bromate, potassium, bromic acid, potassium salt, bromic acid, potassium salt 1:1, ccris 529, eec no. e924, kaliumbromat, potassium bromate, potassium bromate dot, potassiumbromate, unii-04mb35w6za | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
potassium;bromate | |
23673461 | |
167.01 | |
99.5% | |
500ppm max. | |
5ppm max. | |
0.005% max. (in water) | |
0.1% max. (105°C) | |
100ppm max. | |
3.27 | |
0.001% max. (as N) | |
350.0°C | |
1kg |
Chemical Identifiers
7758-01-2 | |
167.01 | |
OCATYIAKPYKMPG-UHFFFAOYSA-M | |
23673461 | |
potassium;bromate |
BrKO3 | |
MFCD00011359 | |
bromate, potassium, bromic acid, potassium salt, bromic acid, potassium salt 1:1, ccris 529, eec no. e924, kaliumbromat, potassium bromate, potassium bromate dot, potassiumbromate, unii-04mb35w6za | |
CHEBI:38211 | |
[O-]Br(=O)=O.[K+] |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
May cause cancer.
May cause fire or explosion; strong oxidizer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 231-829-8
RTECSNumber : EF8725000
TSCA : TSCA