missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium borohydride, 98%, Alfa Aesar™

Source of Hydrogen, Reducing Agent For Aldehydes, Ketones

Manufacturer:  Alfa Aesar 3257009

 View more versions of this product

Catalog No. AA3257009



Specifications

Specifications

Potassium borohydride
98%
MFCD00011396
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
[B-].[K+]
49.908
53.94
1.17g/cm3 at 20°C
White
13762-51-1
BK
White powder
HHUJLKPGQMFFMS-UHFFFAOYSA-N
potassium;boron(1-)
22892188
Powder
Odorless
10g

Chemical Identifiers

13762-51-1
49.908
HHUJLKPGQMFFMS-UHFFFAOYSA-N
22892188
[B-].[K+]
BK
MFCD00011396
potassium borohydride, potassium tetrahydroborate, potassium borohydrate, unii-0ws230dtgf, potassium tetrahydroborate 1-, borohydrure de potassium french, borate 1-, tetrahydro-, potassium, 0ws230dtgf, borohydrure de potassium, borylpotassium
potassium;boron(1-)
Safety and Handling

Safety and Handling

H261-H301-H311-H331-H314-H318: In contact with water releases flammable gas. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.3; Packing Group: I

EINECSNumber : 237-360-5

SDS Documents