Please login to your online account to display your discounted pricing

Potassium Bisulfite, Reagent Grade, LabChem™

$250.80 - $250.80

Chemical Identifiers

Chemical Name or Material Potassium Bisulfite
CAS 16731-55-8
CAS Max % 100
Purity Grade Reagent Grade
Linear Formula K2S2O5
Formula Weight 222.33
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC187701

 
lab chem
LC187701
500g Each for $250.80
Description & Specifications

Specifications

Formula Weight 222.33
Storage Information Room Temperature
EINECS Number 240-795-3
Physical Form Powder
CAS Max % 100
Chemical Name or Material Potassium Bisulfite
CAS 16731-55-8
Health Hazard 2 GHS H Statement
Causes serious eye damage.
Health Hazard 3 GHS P Statement
Wear eye protection.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Color White
Decomposition Information Sulfur compounds; Potassium oxide
Quantity 500g
Identification Passes Test
Solubility Soluble in water
Linear Formula K2S2O5
Packaging Amber Glass
Melting Point 150°C
Purity Grade Reagent Grade
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
Wear eye protection.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.

GHS H Statement
Causes serious eye damage.