Please login to your online account to display your discounted pricing

Potassium Bisulfate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$294.04 - $1,016.79

Chemical Identifiers

Chemical Name or Material Potassium Bisulfate
CAS 7646-93-7
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula KHSO4
Formula Weight 136.17
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003714

 
avantor performance
296005
Wide Mouth Amber Glass Bottle 2.5kg Each for $1,016.79

02003715

 
avantor performance
296001
Poly Bottle 500g Each for $294.04
Description & Specifications

Specifications

Formula Weight 136.17
Physical Form Crystals
Chemical Name or Material Potassium Bisulfate
CAS 7646-93-7
Purity Grade BAKER ANALYZED™ Reagent
Packaging Wide Mouth Amber Glass Bottle
Quantity 2.5kg
Specific Gravity 1L = 2.24kg
Molecular Formula KHSO4