Please login to your online account to display your discounted pricing

Potassium Bisulfate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  296001

 View more versions of this product

Catalog No. 02003715

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 136.17
Chemical Name or Material Potassium Bisulfate
CAS 7646-93-7
Molecular Formula KHSO4
Specific Gravity 1L = 2.24kg
Quantity 500g
Purity Grade BAKER ANALYZED™ Reagent
Packaging Poly Bottle

Specifications

Chemical Name or Material Potassium Bisulfate
CAS 7646-93-7
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula KHSO4
Formula Weight 136.17