Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium bis(trimethylsilyl)amide, 15% w/w in toluene

Chemical Identifiers

Chemical Name or Material Potassium bis(trimethylsilyl)amide
CAS 40949-94-8
Molecular Formula C6H18KNSi2
Linear Formula [(CH3)3Si]2NK
Formula Weight 199.49
MDL Number MFCD00010330
UN Number UN2924
Beilstein 4006754
Synonym KHMDS; Potassium hexamethyldisilazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL15022AP

 
alfa aesar™
L15022AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 199.49
Molecular Formula C6H18KNSi2
Linear Formula [(CH3)3Si]2NK
MDL Number MFCD00010330
CAS 40949-94-8
Beilstein 4006754
Synonym KHMDS; Potassium hexamethyldisilazide
Flash Point 7°C (44°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225-H314-H318-H361-H373-H336
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
May cause drowsiness or dizziness.
Health Hazard 3 GHS P Statement
P210-P280-P301+P310-P305+P351+P338-P315-P402
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Get immediate medical advice/attention.
Store in a dry place.
Concentration 15% w/w in toluene
Chemical Name or Material Potassium bis(trimethylsilyl)amide
UN Number UN2924
Quantity 500mL
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

Applications
Potassium bis(trimethylsilyl)amide is used in the generation of K enolates. It is also used in the Wittig reaction, to give Li salt-free product distribution for selective (Z)-alkene formation and used with 18-crown-6 for selective formation of (Z)-ɑß-unsaturated esters from phosphonates.

Solubility
Reacts with water.

Notes
It is sensitive to moisture and air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Handle and store under inert gas. Incompatible with strong oxidizing agents.