Please login to your online account to display your discounted pricing

Potassium bis(trimethylsilyl)amide, 0.5M solution in toluene, +99.4%, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC418238000

 View more versions of this product

Catalog No. AC418238000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 199.49
Concentration 0.6 to 0.8M (Total base)
Quantity 800mL
Color Clear yellow
Packaging AcroSeal™ Glass Bottle
Assay 0.6 to 0.8M (Total base)
Density 0.8760g/mL
Synonym Potassiumhexamethyldisilazane, 0.5M solution in toluene
Linear Formula [(CH3)3Si]2NK
CAS 108-88-3
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects if inhaled. Harmful to aquatic life with long lasting effects.
Molecular Formula C6H18KNSi2
Health Hazard 1 Danger
CAS 40949-94-8
Health Hazard 3 GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Obtain special instructions before use. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid release to the environment.
Chemical Name or Material Potassium bis(trimethylsilyl)amide
MDL Number MFCD00010330
Name Note 0.5M solution in toluene

Specifications

Chemical Name or Material Potassium bis(trimethylsilyl)amide
Name Note 0.5M solution in toluene
CAS 40949-94-8
CAS 108-88-3
Assay 0.6 to 0.8M (Total base)
Molecular Formula C6H18KNSi2
Linear Formula [(CH3)3Si]2NK
Formula Weight 199.49
MDL Number MFCD00010330
Synonym Potassiumhexamethyldisilazane, 0.5M solution in toluene
Accessories

Danger

GHS P Statement IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Obtain special instructions before use. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Avoid release to the environment.

GHS P Statement Causes severe skin burns and eye damage. May be fatal if swallowed and enters airways. May cause drowsiness or dizziness. May cause damage to organs through prolonged or repeated exposure. Suspected of damaging the unborn child. Highly flammable liquid and vapor. Suspected of causing cancer. Suspected of causing genetic defects if inhaled. Harmful to aquatic life with long lasting effects.

SDS & Safety Documents