missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 46909MD

 View more versions of this product

Catalog No. AA46909MD



Specifications

Specifications

Potassium bis (trifluoromethylsulfonyl)imide
C2F6KNO4S2
MFCD06200829
Potassium triflimide; Potassium trifluoromethanesulfonimide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+]
319.234
319.24
198°C to 203°C
Moisture Sensitive
90076-67-8
(CF3SO2)2NK
UN1759
KVFIZLDWRFTUEM-UHFFFAOYSA-N
potassium;bis(trifluoromethylsulfonyl)azanide
11099217
Solid
250mg

Chemical Identifiers

90076-67-8
319.234
KVFIZLDWRFTUEM-UHFFFAOYSA-N
11099217
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+]
C2F6KNO4S2
MFCD06200829
Potassium triflimide; Potassium trifluoromethanesulfonimide
potassium;bis(trifluoromethylsulfonyl)azanide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

SDS Documents