Learn More
Potassium bis(trifluoromethylsulfonyl)imide, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04690906
Description
Potassium bis(trifluoromethylsulfonyl)imide is used as Flavor & Fragrance intermediates. It is used as an intermediate in Industries such as agrochemical, dyestuff, pharmaceutical, syntheses material, chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium bis (trifluoromethylsulfonyl)imide | |
198°C to 203°C | |
(CF3SO2)2NK | |
5 g | |
Moisture sensitive | |
It is soluble in water. 10mg/mL. | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+] | |
319.234 | |
319.24 |
90076-67-8 | |
C2F6KNO4S2 | |
MFCD06200829 | |
UN1759 | |
potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide | |
KVFIZLDWRFTUEM-UHFFFAOYSA-N | |
potassium;bis(trifluoromethylsulfonyl)azanide | |
11099217 | |
Solid |
Chemical Identifiers
90076-67-8 | |
319.234 | |
KVFIZLDWRFTUEM-UHFFFAOYSA-N | |
11099217 | |
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[K+] |
C2F6KNO4S2 | |
MFCD06200829 | |
potassium bistriflylimide anion, methanesulfonamide,1,1,1-trifluoro-n-trifluoromethyl sulfonyl-, potassium salt 1:1, potassium bis trifluoromethanesulfonly imide, ktfsi, potassium trifluoromethanesulfonimide, potassiobis trifluoromethylsulfonyl amine, potassium bis trifluoromethanesulfonyl amide, potassium bis trifluoromethyl sulfonyl imide, 1,1,1-trifluoro-n-potassio-n-trifluoromethanesulfonylmethanesulfonamide | |
potassium;bis(trifluoromethylsulfonyl)azanide |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only