Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.997% (metals basis)

Chemical Identifiers

Chemical Name or Material Potassium bis (oxalato)oxotitanate (IV) hydrate
CAS 14402-67-6
Purity Grade Puratronic™
Molecular Formula C4K2O9Ti·xH2O
Linear Formula K2[TiO(C2O4)2]·xH2O
Formula Weight 318.10 (Anhydrous)
MDL Number MFCD00150592
UN Number UN3288
Synonym Potassium titanyloxolate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4289804

  SDS
alfa aesar™
4289804
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4289809

  SDS
alfa aesar™
4289809
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4289818

  SDS
alfa aesar™
4289818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Potassium bis (oxalato)oxotitanate (IV) hydrate
CAS 14402-67-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
TSCA TSCA
Purity Grade Puratronic™
Physical Form Coarse Crystalline Powder
Assay 99.997%
Assay Percent Notes (metals basis)
Molecular Formula C4K2O9Ti·xH2O
Linear Formula K2[TiO(C2O4)2]·xH2O
Formula Weight 318.10 (Anhydrous)
MDL Number MFCD00150592
UN Number UN3288
Synonym Potassium titanyloxolate
Odor Odorless
Color Colorless
Quantity 2g
Safety and Handling

Warning

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.