missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Biiodate, For Residual Chlorine, Certified, 0.0282N ±0.0005N (0.00235M), LabChem™

$68.91 - $68.91

Chemical Identifiers

CAS 7732-18-5,13455-24-8
CAS Max % 99.91,0.09
Linear Formula KHI2O6
Formula Weight 389.92
Chemical Name or Material Potassium Biiodate, For Residual Chlorine
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC187351 Lab Chem
LC187351
500mL Each for $68.91
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,13455-24-8
KHI2O6
Potassium Biiodate, For Residual Chlorine
99.91,0.09
389.92
Certified Grade

Specifications

389.92
Iodine vapor
0.09,99.91
Poly Bottle
KHI2O6
Room Temperature
0.0282N ±0.0005N (0.00235M)
Clear, colorless
Potassium Biiodate, For Residual Chlorine
13455-24-8,7732-18-5
1g/mL
500mL
Soluble in water
1g/mL
Passes Test
Liquid
Traceable to NIST
Certified Grade

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.