missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Biiodate, ACS Grade, 99.95 to 100.05%, LabChem™

$223.94 - $223.94

Chemical Identifiers

CAS 13455-24-8
CAS Max % 100
Linear Formula KHI2O6
Formula Weight 389.92
UN Number UN3085
Chemical Name or Material Potassium Biiodate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC187259 Lab Chem
LC187259
100g Each for $223.94
Specifications Description & Specifications

Chemical Identifiers

13455-24-8
KHI2O6
UN3085
ACS Grade
100
389.92
Potassium Biiodate

Specifications

389.92
White
13455-24-8
Room Temperature
100
Iodine vapor
KHI2O6
99.95 to 100.05%, by acid-base titrimetry
100g
Powder
Soluble in water
UN3085
Passes Test
Amber Glass
Potassium Biiodate
ACS Grade

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat, sparks, open flames, hot surfaces.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Do not breathe dust.
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 236-650-9