missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Bicarbonate (Powder/USP/FCC), Fisher Chemical

Manufacturer:  Thermo Scientific FLP235500

 View more versions of this product

Catalog No. P235-500




Specifications

Specifications

Potassium Bicarbonate
FCC/USP
(Dried Basis)99.5to101.5%
CHKO3
Potassium Acid Carbonate, Potassium Hydrogen Carbonate
C(=O)(O)[O-].[K+]
100.114
CHEBI:81862
Powder/Solid
0.001% max. (USP), 0.001% max. (USP), 10mg/kg max. (FCC), 10mg/kg max. (FCC)
2mg/kg max. (FCC)
2.5% max. (USP), 2.5% max. (USP), Pass Test (FCC), Pass Test (FCC)
Meets Requirements (USP)
White
500g
298-14-6
99.5 to 101.5% (USP)
(Dried Basis)99.5-101.5%
MFCD00011402
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium;hydrogen carbonate
516893
100.12
2.17g/cm³
Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC)
0.25% max. (FCC), 0.3% max. (USP), 0.3% max. (USP), 0.25% max. (FCC)
Poly Bottle
>100°C
100°C

Chemical Identifiers

298-14-6
100.114
TYJJADVDDVDEDZ-UHFFFAOYSA-M
516893
CHKO3
MFCD00011402
Potassium Acid Carbonate, Potassium Hydrogen Carbonate
CHEBI:81862
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May be harmful by inhalation, in contact with skin and if swallowed. Wear personal protective equipment. Use personal protective equipment. Look out! DATATYPE 710. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. . . . .

NFPA

Health:1
Flammability:0
Instability:0

SDS Documents