Please login to your online account to display your discounted pricing

Potassium Bicarbonate (Powder/USP/FCC), Fisher Chemical

$290.93 - $3,151.29

Chemical Identifiers

Chemical Name or Material Potassium Bicarbonate
CAS 298-14-6
Purity Grade FCC/USP
Physical Form Powder/Solid
Assay 99.0 to 101.5% (FCC)
Assay Percent Range (Dried Basis)99.5to101.5%
Assay Percent Notes (Dried Basis)99.5-101.5%
Molecular Formula KHCO3
Formula Weight 100.12
MDL Number MFCD00011402
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

P235-12

  SDS
fisher scientific company
FLP23512
Poly Pail 12kg Each for $3,151.29

P235-212

  SDS
fisher scientific company
FLP235212
Poly Bottle 2.5kg Each for $593.50
Case of 4 Each for $2,374.00

P235-500

  SDS
fisher scientific company
FLP235500
Poly Bottle 500g Each for $290.93
Case of 6 Each for $1,745.58
Description & Specifications

Specifications

Purity Grade FCC/USP
Physical Form Powder/Solid
Formula Weight 100.12
Melting Point 100°C
Quantity 12kg
Assay 99.0 to 101.5% (FCC)
Viscosity [NULL]
Decomposition Information >100°C
Color White
Density 2.17g/cm³
Identification Pass Test (FCC)
Loss on Drying 0.25% max. (FCC)
Residual Solvents Meets Requirements (USP)
Synonym Potassium Acid Carbonate
Health Hazard 2 CAUTION!
Chemical Name or Material Potassium Bicarbonate
Heavy Metals (as Pb) 0.001% max. (USP)
Assay Percent Notes (Dried Basis)99.5-101.5%
Health Hazard 3 Emergency Overview
May be harmful by inhalation, in contact with skin and if swallowed. Wear personal protective equipment. Use personal protective equipment. Look out! DATATYPE 710. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. . . . .

NFPA

Health:1
Flammability:0
Instability:0
Normal Carbonate 2.5% max. (USP)
MDL Number MFCD00011402
Lead (Pb) 2mg/kg max. (FCC)
Packaging Poly Pail
Assay Percent Range (Dried Basis)99.5to101.5%
Molecular Formula KHCO3
CAS 298-14-6
ChemAlert Storage Symbol Gray
Safety and Handling
 

Emergency Overview
May be harmful by inhalation, in contact with skin and if swallowed. Wear personal protective equipment. Use personal protective equipment. Look out! DATATYPE 710. Avoid contact with skin, eyes and clothing. Avoid ingestion and inhalation. . . . .

NFPA

Health:1
Flammability:0
Instability:0

CAUTION!