missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Bicarbonate ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 298-14-6
Physical Form Granular
Molecular Formula KHCO3
Formula Weight 100.12
Chemical Name or Material Potassium Bicarbonate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK6748212 Avantor Performance
674806
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
MK6748500 Avantor Performance
674803
POLYSTORMOR 500g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

298-14-6
KHCO3
Potassium Bicarbonate
Granular
100.12
ACS Grade

Specifications

298-14-6
KHCO3
Poly
Potassium Bicarbonate
Granular
100.12
2.5kg
ACS Grade