Please login to your online account to display your discounted pricing

Potassium Bicarbonate ACS AR Granular, Macron Fine Chemicals™


macron

Quantity 500g
Packaging POLYSTORMOR

Manufacturer: avantor performance  674803

 View more versions of this product

Catalog No. MK6748500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 100.12
Purity Grade ACS Grade
Physical Form Granular
Quantity 500g
Packaging POLYSTORMOR
Molecular Formula KHCO3
CAS 298-14-6
Chemical Name or Material Potassium Bicarbonate

Specifications

Chemical Name or Material Potassium Bicarbonate
CAS 298-14-6
Purity Grade ACS Grade
Physical Form Granular
Molecular Formula KHCO3
Formula Weight 100.12