missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium benzoate, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1161030

 View more versions of this product

Catalog No. AAA1161030



Description

Description

Potassium Benzoate is manufactured primarily for food and beverage use. It is a chemical preservative, which in very low concentrations inhibits the activity of the microorganisms. It is used in carbonated beverages. The shelf life of un-pasteurized cider can be greatly extended by adding potassium benzoate. It is also used as the whistle in many fireworks.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium Benzoate is manufactured primarily for food and beverage use. It is a chemical preservative, which in very low concentrations inhibits the activity of the microorganisms. It is used in carbonated beverages. The shelf life of un-pasteurized cider can be greatly extended by adding potassium benzoate. It is also used as the whistle in many fireworks.

Solubility
Soluble in ethanol, slightly soluble in methanol, insoluble in ether.

Notes
Hygroscopic. Protect from humidity and water. Store under dry inert gas away from oxidizing agents and moisture.
Specifications

Specifications

Potassium Benzoate
>300°C
C7H5KO2
250 g
Hygroscopic
potassium benzoate, benzoic acid, potassium salt, potassium salt, unii-763yqn2k7k, potassium benzoate nf, benzoic acid potassium salt, potassium ion benzoate, acmc-20ajv6, dsstox_cid_7219
XAEFZNCEHLXOMS-UHFFFAOYSA-M
potassium;benzoate
23661960
99%
582-25-2
99%
MFCD00013061
3719165
14,1091
Soluble in ethanol,slightly soluble in methanol,insoluble in ether.
C1=CC=C(C=C1)C(=O)[O-].[K+]
160.213
160.22

Chemical Identifiers

582-25-2
160.213
XAEFZNCEHLXOMS-UHFFFAOYSA-M
23661960
C1=CC=C(C=C1)C(=O)[O-].[K+]
C7H5KO2
MFCD00013061
potassium benzoate, benzoic acid, potassium salt, potassium salt, unii-763yqn2k7k, potassium benzoate nf, benzoic acid potassium salt, potassium ion benzoate, acmc-20ajv6, dsstox_cid_7219
potassium;benzoate
Safety and Handling

Safety and Handling

EINECSNumber : 209-481-3

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only