Learn More
Potassium antimonyl tartrate sesquihydrate, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 223805000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium antimonyl tartrate sesquihydrate | |
99+% | |
28300-74-5 | |
500 g | |
1·5 H2O | |
15, 691 | |
Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water, 1g/15mL glycerol, insoluble in alcohol | |
333.93 | |
Authentic | |
Glass bottle |
100.0°C | |
White | |
Fine Crystalline Powder | |
99% min. (Iodometry) | |
MFCD00167056 | |
Antimonyl potassium tartrate sesquihydrate | |
333.93 | |
≥99% | |
2.7% max. (105°C) |
Chemical Identifiers
28300-74-5 | |
333.93 | |
Antimonyl potassium tartrate sesquihydrate |
1·5 H2O | |
MFCD00167056 |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
GHS Signal Word: Warning
RTECSNumber : CC6825000
RUO – Research Use Only