Please login to your online account to display your discounted pricing

Potassium Antimony Tartrate, ACS Grade, 99.0 to 103.0%, LabChem™

$132.03 - $132.03

Chemical Identifiers

Chemical Name or Material Potassium Antimony Tartrate
CAS 28300-74-5
CAS Max % 100
Purity Grade ACS Grade
Linear Formula C8H4K2O12Sb2·3H2O
Formula Weight 667.87
UN Number UN1551
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC187109

 
lab chem
LC187109
100g Each for $132.03
Description & Specifications

Specifications

Formula Weight 667.87
Quantity 100g
Acidity Alkalinity <0.020meq/gcSt
Chemical Name or Material Potassium Antimony Tartrate
EINECS Number 234-293-3
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Powder
Assay 99.0 to 103.0%
Solubility Soluble in water
Specific Gravity 2600kg/m3
Storage Information Room Temperature
Linear Formula C8H4K2O12Sb2·3H2O
CAS 28300-74-5
UN Number UN1551
Decomposition Information Carbon monoxide; Carbon dioxide; Antimony and its oxides
Color White
Health Hazard 1 Danger
CAS Max % 100
Health Hazard 2 GHS H Statement
Toxic if swallowed.
Toxic to aquatic life.
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Density 2,600kg/m3
Safety and Handling

Danger

GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Toxic if swallowed.
Toxic to aquatic life.