missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Allyltrifluoroborate, TCI America™

Manufacturer:  TCI America P14805G

 View more versions of this product

Catalog No. P14805G



Specifications

Specifications

Potassium Allyltrifluoroborate
5g
C3H5BF3K
[B-](CC=C)(F)(F)F.[K+]
147.977
147.98
≥97.0% (W)
White
233664-53-4
TVPZAMJXLCDMIT-UHFFFAOYSA-N
potassium;trifluoro(prop-2-enyl)boranuide
23664277
Crystal-Powder at 20°C

Chemical Identifiers

233664-53-4
147.977
23664277
[B-](CC=C)(F)(F)F.[K+]
C3H5BF3K
TVPZAMJXLCDMIT-UHFFFAOYSA-N
potassium;trifluoro(prop-2-enyl)boranuide
Safety and Handling

Safety and Handling

TSCA : Yes

Documents