Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium acrylate hydrate, 95%

$70.95 - $303.24

Chemical Identifiers

Chemical Name or Material Potassium acrylate hydrate
CAS 10192-85-5
Molecular Formula H2C=CHCO2K·xH2O
Formula Weight 110.16 (Anhydrous)
MDL Number MFCD00151663
Beilstein 3695913
Synonym Acrylic acid potassium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1007906

 
alfa aesar™
L1007906
5g Each for $70.95

AAL1007914

 
alfa aesar™
L1007914
25g Each for $303.24
Description & Specifications

Specifications

Formula Weight 110.16 (Anhydrous)
Chemical Name or Material Potassium acrylate hydrate
Assay 95%
MDL Number MFCD00151663
TSCA TSCA
Molecular Formula H2C=CHCO2K·xH2O
CAS 10192-85-5
Beilstein 3695913
Synonym Acrylic acid potassium salt
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Melting Point >300°C
Quantity 5g
Health Hazard 1 Warning
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C