missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Acid Phthalate, Certified, 0.0500N ±0.0005N (0.05M), LabChem™

Manufacturer:  LabChem LC187002

 View more versions of this product

Catalog No. LC187002



Specifications

Specifications

Potassium Acid Phthalate
1g/mL
Poly Bottle
Colorless
877-24-7,7732-18-5
C8H5KO4
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
Soluble in water
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
204.222
204.23
Certified
Carbon monoxide; Carbon dioxide
Passes Test
1g/mL
1L
98.98,1.02
KHC8H4O4
0.0500N ±0.0005N (0.05M)
IWZKICVEHNUQTL-UHFFFAOYSA-M
potassium;2-carboxybenzoate
23676735
Liquid

Chemical Identifiers

877-24-7
204.222
potassium hydrogen phthalate, potassium biphthalate, monopotassium phthalate, potassium acid phthalate, hydrogen potassium phthalate, phthalic acid monopotassium salt, 1,2-benzenedicarboxylic acid, monopotassium salt, phthalic acid, monopotassium salt, phthalic acid potassium salt, potassium phthalate monobasic
potassium;2-carboxybenzoate
C8H5KO4
IWZKICVEHNUQTL-UHFFFAOYSA-M
23676735
C1=CC=C(C(=C1)C(=O)O)C(=O)[O-].[K+]
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

Documents
Promotions

Promotions