Please login to your online account to display your discounted pricing

Potassium Acid Phthalate, ACS Grade, 99.95 to 100.05%, LabChem™

$185.46 - $185.46

Chemical Identifiers

Chemical Name or Material Potassium Acid Phthalate
CAS 877-24-7
CAS Max % 100
Purity Grade ACS Grade
Linear Formula KHC8H4O4
Formula Weight 204.23
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC186901

 
lab chem
LC186901
Poly Bottle 500g Each for $185.46
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 204.23
Chemical Name or Material Potassium Acid Phthalate
EINECS Number 212-889-4
Purity Grade ACS Grade
Identification Passes Test
Assay 99.95 to 100.05%
Linear Formula KHC8H4O4
CAS 877-24-7
Packaging Poly Bottle
Health Hazard 1 Warning
Solubility Soluble in water
Specific Gravity 1.64g/cm3
CAS Max % 100
Health Hazard 2 GHS H Statement
Causes eye irritation.
Storage Information Room Temperature
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Decomposition Information Carbon monoxide; Carbon dioxide
Density 1.64g/cm3
Color White
Quantity 500g
Safety and Handling

Warning

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes eye irritation.