Please login to your online account to display your discounted pricing

Potassium Acid Phthalate, 0.0500N , Ricca Chemical

$99.61 - $159.52

Chemical Identifiers

Chemical Name or Material Potassium Acid Phthalate
CAS 7732-18-5 (Water, Deionized)
CAS 877-24-7 (Potassium Acid Phthalate)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5870-16

 
ricca chemical company
RI587016
Plastic 500mL Each for $99.61

5870-32

 
ricca chemical company
RI587032
Plastic 1L Each for $159.52
Description & Specifications

Specifications

Quantity 500mL
Methods APHA for Acidity
Chemical Name or Material Potassium Acid Phthalate
Packaging Plastic
CAS 7732-18-5 (Water, Deionized)
Concentration or Composition (by Analyte or Components) 0.0500N (N/20)
CAS 877-24-7 (Potassium Acid Phthalate)