missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Acid Phthalate, 0.0500 N (N/20), Ricca Chemical
For Precision Titrations Testing
Supplier: Ricca Chemical Company 587016
Specifications
Potassium Acid Phthalate | |
Natural Poly Bottle | |
APHA for Acidity | |
500 mL |