Please login to your online account to display your discounted pricing

Potassium Acetate TS, 10% (w/v) Aqueous Solution, Ricca Chemical

$137.39 - $137.39

Chemical Identifiers

Chemical Name or Material Potassium Acetate
Name Note Test Solution
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 127-08-2
CAS Min % 8.91
CAS Max % 9.28
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5866-4

 
ricca chemical company
RI58664
Poly natural 120mL Each for $137.39

5866-16

 
ricca chemical company
RI586616
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 120mL
Concentration or Composition Notes 10% (w/v)
Solvent or Matrix Aqueous Solution
CAS Min % 89.0
CAS Max % 93.0
Chemical Name or Material Potassium Acetate
Name Note Test Solution
CAS 7732-18-5
CAS 127-08-2
Packaging Poly natural
CAS Min % 8.91
CAS Max % 9.28