missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Acetate ACS AR Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 127-08-2
Physical Form Crystalline
Molecular Formula CH3COOK
Formula Weight 98.14
Chemical Name or Material Potassium Acetate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK6700212 Avantor Performance
670012
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

127-08-2
CH3COOK
Potassium Acetate
Crystalline
98.14
ACS Grade

Specifications

127-08-2
CH3COOK
Poly
Potassium Acetate
Crystalline
98.14
2.5kg
ACS Grade