Please login to your online account to display your discounted pricing

Potassium acetate, ACS, 99% min, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Potassium acetate
CAS 127-08-2
Purity Grade ACS Grade
Molecular Formula C2H3KO2
Linear Formula CH3CO2K
Formula Weight 98.15
MDL Number MFCD00012458
Beilstein 3595449
Merck Index 14,7605
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA13449A3

 
alfa aesar
13449A3
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 98.15
Quantity 2kg
Chemical Name or Material Potassium acetate
TSCA TSCA
Physical Form Crystalline
Assay ≥99%
Sensitivity Hygroscopic
Molecular Formula C2H3KO2
Linear Formula CH3CO2K
MDL Number MFCD00012458
CAS 127-08-2
Beilstein 3595449
Merck Index 14,7605
Color White
Melting Point 292°C
Solubility Information Soluble in water and alcohol
Density 1.57
RTECS Number AJ3325000