missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium acetate, ACS, 99% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 13449A3

 View more versions of this product

Catalog No. AA13449A3



Specifications

Specifications

Potassium acetate
C2H3KO2
MFCD00012458
14,7605
Soluble in water and alcohol
CC(=O)[O-].[K+]
98.142
CHEBI:32029
Crystalline
ACS
Hygroscopic
292°C
127-08-2
CH3CO2K
3595449
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.15
≥99%
1.57
White
2kg

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
MFCD00012458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]
Safety and Handling

Safety and Handling

RTECSNumber : AJ3325000

TSCA : TSCA

Documents