missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium acetate, 8M aq. soln., RNAse free

$95.46 - $95.46

Chemical Identifiers

CAS 127-08-2
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.142
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
PubChem CID 517044
ChEBI CHEBI:32029
IUPAC Name potassium;acetate
SMILES CC(=O)[O-].[K+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62856AK View Documents Alfa Aesar™
J62856AK
250mL Each for $95.46
 
Description

Description

Applications
Potassium Acetate is a potassium salt employed to replenish electrolytes, for restoration of water-electrolyte balance, as well as a urinary and systemic alkalizer. Potassium Acetate can be administered orally or by intravenous infusion. Formerly, It was employed in diuretics and expectorants. A potassium source employed in DNA and protein purification. Some other applications includes as buffering agent, neutralizer, preservative, color retention agent(protect natural color of animals and plants).

Solubility
Soluble in water (98.2 g/L completely soluble) at 25°C, alcohol, methanol, and ammonia. Insoluble in acetone, and ether.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
MFCD00012458
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CHEBI:32029
CC(=O)[O-].[K+]

Specifications

127-08-2
CH3CO2K
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920
CC(=O)[O-].[K+]
98.142
CHEBI:32029
Liquid
Colorless
Potassium acetate, RNAse free
C2H3KO2
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.15
8M aq. soln.
250mL
Safety and Handling

Safety and Handling

TSCA : TSCA