missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium acetate, 1M aq. soln., pH 7.5

Manufacturer:  Alfa Aesar™ J62817AE

 View more versions of this product

Catalog No. AAJ62817AE



Specifications

Specifications

Potassium acetate
C2H3KO2
MFCD00012458
acetic acid potassium salt, acetic acid, potassium salt, acok, diuretic salt, fema no. 2920, koac, octan draselny czech, potassium acetate, potassium ethanoate, potassiumacetate
CC(=O)[O-].[K+]
98.142
CHEBI:32029
Liquid
7.5
127-08-2
CH3CO2K
1M aq. soln.
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium;acetate
517044
98.15
Colorless
100mL

Chemical Identifiers

127-08-2
98.142
SCVFZCLFOSHCOH-UHFFFAOYSA-M
517044
potassium;acetate
C2H3KO2
MFCD00012458
acetic acid potassium salt, acetic acid, potassium salt, acok, diuretic salt, fema no. 2920, koac, octan draselny czech, potassium acetate, potassium ethanoate, potassiumacetate
CHEBI:32029
CC(=O)[O-].[K+]
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents