Please login to your online account to display your discounted pricing

Potassium AA Standard, Certified, 1000ppm K ±10 (1mL = 0.1mg K), LabChem™


Quantity 500mL
Packaging Poly Bottle

Manufacturer: lab chem  LC201101

 View more versions of this product

Catalog No. LC201101

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 74.55
Chemical Name or Material Potassium AA Standard
Physical Form Liquid
Solubility Soluble in water
CAS Max % 99.98
Traceability to NIST Traceable to NIST
Concentration 1000ppm K ±10 (1mL = 0.1mg K)
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Quantity 500mL
Linear Formula KCl
Density 1g/mL
Packaging Poly Bottle
CAS 7732-18-5
CAS Max % 0.02
Color Clear, colorless
CAS 7447-40-7
Recommended Storage Room Temperature
Name Note 100ppm (1mL = 0.1mg K)
Specific Gravity 1g/mL
Identification Passes Test
Decomposition Information Hydrogen chloride; Potassium oxide

Specifications

Chemical Name or Material Potassium AA Standard
CAS 7732-18-5
CAS Max % 99.98
CAS 7447-40-7
CAS Max % 0.02
Name Note 100ppm (1mL = 0.1mg K)
Purity Grade Certified Grade
Linear Formula KCl
Formula Weight 74.55
Accessories

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature