missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium AA Standard, Certified, 1000ppm K ±10 (1mL = 0.1mg K), LabChem™

$62.89 - $62.89

Chemical Identifiers

CAS 7732-18-5,7447-40-7
CAS Max % 99.98,0.02
Linear Formula KCl
Formula Weight 74.55
Chemical Name or Material Potassium AA Standard, 100ppm (1mL = 0.1mg K)
Grade Certified Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
LC201101 Lab Chem
LC201101
Poly Bottle 500mL Each for $62.89
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7447-40-7
KCl
Potassium AA Standard, 100ppm (1mL = 0.1mg K)
99.98,0.02
74.55
Certified Grade

Specifications

7732-18-5,7447-40-7
Liquid
KCl
Passes Test
Soluble in water
1g/mL
500mL
Certified Grade
99.98,0.02
1000ppm K ±10 (1mL = 0.1mg K)
74.55
Poly Bottle
Traceable to NIST
Clear, colorless
Potassium AA Standard, 100ppm (1mL = 0.1mg K)

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature