missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium 5-Methyl-1,3,4-oxadiazole-2-carboxylate, TCI America™

Manufacturer:  TCI America P2220200MG

 View more versions of this product

Catalog No. P2220200MG



Specifications

Specifications

Potassium 5-Methyl-1,3,4-oxadiazole-2-carboxylate
200mg
C4H3KN2O3
5-Methyl-1,3,4-oxadiazole-2-carboxylic Acid Potassium Salt
CC1=NN=C(O1)C(=O)[O-].[K+]
166.177
166.18
≥98.0% (HPLC,T)
White-Yellow
888504-28-7
MFCD08060085
BRHMYHXNUGVBCU-UHFFFAOYSA-M
potassium;5-methyl-1,3,4-oxadiazole-2-carboxylate
24820502
Crystal-Powder at 20°C

Chemical Identifiers

888504-28-7
166.177
BRHMYHXNUGVBCU-UHFFFAOYSA-M
24820502
CC1=NN=C(O1)C(=O)[O-].[K+]
C4H3KN2O3
MFCD08060085
5-Methyl-1,3,4-oxadiazole-2-carboxylic Acid Potassium Salt
potassium;5-methyl-1,3,4-oxadiazole-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Documents