Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 5-formylthiophene-2-trifluoroborate, 97%

$235.27 - $235.27

Chemical Identifiers

Chemical Name or Material Potassium 5-formylthiophene-2-trifluoroborate
CAS 1025113-78-3
Molecular Formula C5H3BF3KOS
Formula Weight 218.05g/mol
MDL Number MFCD09800740
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2810206

 
alfa aesar™
H28102-06
5g Each for $235.27
Description & Specifications

Specifications

Melting Point 245°C
Quantity 5g
Chemical Name or Material Potassium 5-formylthiophene-2-trifluoroborate
CAS 1025113-78-3
Assay 97%
Molecular Formula C5H3BF3KOS
Formula Weight 218.05g/mol
MDL Number MFCD09800740
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.