Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 5-formylthiophene-2-trifluoroborate

$79.05 - $302.00

Chemical Identifiers

Chemical Name or Material Potassium 5-formylthiophene-2-trifluoroborate
CAS 1025113-78-3
Molecular Formula C5H3BF3KOS
Formula Weight 218.05g/mol
MDL Number MFCD09800740
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5114003

 
alfa aesar™
H51140-03
Each for $79.05

AAH5114006

 
alfa aesar™
H51140-06
Each for $302.00
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material Potassium 5-formylthiophene-2-trifluoroborate
CAS 1025113-78-3
Molecular Formula C5H3BF3KOS
Formula Weight 218.05g/mol
MDL Number MFCD09800740
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.