missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-(trifluoromethyl)phenyltrifluoroborate, 96%

Manufacturer:  Alfa Aesar™ H5325906

 View more versions of this product

Catalog No. AAH5325906



Description

Description

Applications
Reactant in the Suzuki-Miyaura Cross-Coupling of Aryl Imidazolylsulfonates in water. Reactant in cross-coupling reaction of potassium aryltrifluoroborates with organic chlorides in aqueous media catalyzed by an oxime-derived palladacycle.

Solubility
High air and moisture stability. Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

Potassium 4-(trifluoromethyl)phenyltrifluoroborate
5g
C7H4BF6K
potassium 4-trifluoromethyl phenyltrifluoroborate, potassium trifluoro 4-trifluoromethyl phenyl borate, potassium trifluoro 4-trifluoromethyl phenyl boranuide, pubchem11514, potassium 4-trifluoromethylphenyl trifluoroborate, potassium 4-trifluoromethylphenyl trifluoroborate,, potassium 4-trifluoromethyl phenyl trifluoroborate, potassium trifluoro-4-trifluoromethyl phenyl boranuide, potassium trifluoro 4-trifluoromethyl phenyl borate 1-, potassium ion trifluoro 4-trifluoromethyl phenyl boranuide
[B-](C1=CC=C(C=C1)C(F)(F)F)(F)(F)F.[K+]
252.008
252.01
280°C to 286°C
166328-08-1
MFCD04039955
KVDSXGFQLXKOID-UHFFFAOYSA-N
potassium;trifluoro-[4-(trifluoromethyl)phenyl]boranuide
23679121
96%

Chemical Identifiers

166328-08-1
252.008
KVDSXGFQLXKOID-UHFFFAOYSA-N
23679121
[B-](C1=CC=C(C=C1)C(F)(F)F)(F)(F)F.[K+]
C7H4BF6K
MFCD04039955
potassium 4-trifluoromethyl phenyltrifluoroborate, potassium trifluoro 4-trifluoromethyl phenyl borate, potassium trifluoro 4-trifluoromethyl phenyl boranuide, pubchem11514, potassium 4-trifluoromethylphenyl trifluoroborate, potassium 4-trifluoromethylphenyl trifluoroborate,, potassium 4-trifluoromethyl phenyl trifluoroborate, potassium trifluoro-4-trifluoromethyl phenyl boranuide, potassium trifluoro 4-trifluoromethyl phenyl borate 1-, potassium ion trifluoro 4-trifluoromethyl phenyl boranuide
potassium;trifluoro-[4-(trifluoromethyl)phenyl]boranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents