Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-(phenylaminocarbonyl)phenyltrifluoroborate, 97%

$43.84 - $165.74

Chemical Identifiers

Chemical Name or Material Potassium 4-(phenylaminocarbonyl)phenyltrifluoroborate
CAS 912350-01-7
Molecular Formula C13H10BF3KNO
Formula Weight 303.13
MDL Number MFCD11977719
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5115003

  SDS
alfa aesar™
H5115003
1g Each for $43.84

AAH5115006

  SDS
alfa aesar™
H5115006
5g Each for $165.74
Description & Specifications

Specifications

Formula Weight 303.13
Quantity 1g
Melting Point >250°C
Assay 97%
Health Hazard 1 Warning
Molecular Formula C13H10BF3KNO
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 912350-01-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Potassium 4-(phenylaminocarbonyl)phenyltrifluoroborate
MDL Number MFCD11977719
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.