Learn More
Alfa Aesar™ Potassium 4-n-butoxyphenyltrifluoroborate, 96%
Manufacturer: Alfa Aesar™ H5309203
Specifications
Potassium 4-n-butoxyphenyltrifluoroborate | |
1g | |
C10H13BF3KO | |
potassium 4-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyltrifluorate, potassium 4-butoxyphenyl trifluoroboranuide, potassium 4-n-butoxyphenyl trifluorate, potassium 4-n-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyl-trifluoroboranuide, potassium 4-butoxyphenyl trifluoro borate 1-, potassium 4-n-butoxyphenyltrifluoroborate, potassium ion 4-butoxyphenyl trifluoroboranuide, potassium 4-butoxyphenyl-tris fluoranyl boranuide | |
[B-](C1=CC=C(C=C1)OCCCC)(F)(F)F.[K+] | |
256.117 | |
256.12 |
>300°C | |
850623-61-9 | |
MFCD04115755 | |
UEVIEKRZQJRPGE-UHFFFAOYSA-N | |
potassium;(4-butoxyphenyl)-trifluoroboranuide | |
44717219 | |
96% |
Chemical Identifiers
850623-61-9 | |
256.117 | |
UEVIEKRZQJRPGE-UHFFFAOYSA-N | |
44717219 | |
[B-](C1=CC=C(C=C1)OCCCC)(F)(F)F.[K+] |
C10H13BF3KO | |
MFCD04115755 | |
potassium 4-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyltrifluorate, potassium 4-butoxyphenyl trifluoroboranuide, potassium 4-n-butoxyphenyl trifluorate, potassium 4-n-butoxyphenyl trifluoroborate, potassium 4-butoxyphenyl-trifluoroboranuide, potassium 4-butoxyphenyl trifluoro borate 1-, potassium 4-n-butoxyphenyltrifluoroborate, potassium ion 4-butoxyphenyl trifluoroboranuide, potassium 4-butoxyphenyl-tris fluoranyl boranuide | |
potassium;(4-butoxyphenyl)-trifluoroboranuide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning