missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-methyl-beta-styryltrifluoroborate, 95%

$84.81 - $283.23

Chemical Identifiers

CAS 219718-86-2
Molecular Formula C9H9BF3K
Formula Weight 224.08
MDL Number MFCD02093982
Beilstein 7783635
Synonym 4-Methyl-beta-styryltrifluoroboric acid potassium salt
Chemical Name or Material Potassium 4-methyl-beta-styryltrifluoroborate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1797303 SDS Alfa Aesar™
L1797303
1g Each for $84.81
AAL1797306 SDS Alfa Aesar™
L1797306
5g Each for $283.23
Specifications Description & Specifications

Chemical Identifiers

219718-86-2
224.08
7783635
Potassium 4-methyl-beta-styryltrifluoroborate
C9H9BF3K
MFCD02093982
4-Methyl-beta-styryltrifluoroboric acid potassium salt

Specifications

219718-86-2
224.08
7783635
1g
95%
C9H9BF3K
MFCD02093982
4-Methyl-beta-styryltrifluoroboric acid potassium salt
Potassium 4-methyl-beta-styryltrifluoroborate

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning