missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium 4-methoxyphenyltrifluoroborate 97%, ACROS Organics™

$78.27 - $78.27

Chemical Identifiers

CAS 192863-36-8
Assay Percent Range 97%
Molecular Formula C7H7BF3KO
Formula Weight 214.04
MDL Number MFCD04112717
Chemical Name or Material Potassium 4-methoxyphenyltrifluoroborate
Assay ≥96.0%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC392910010 SDS Thermo Fisher Scientific
AC392910010
Glass bottle 1g Each for $78.27
Specifications Description & Specifications

Chemical Identifiers

192863-36-8
C7H7BF3KO
MFCD04112717
≥96.0%
97%
214.04
Potassium 4-methoxyphenyltrifluoroborate

Specifications

Powder
1g
192863-36-8
97%
Glass bottle
Potassium 4-methoxyphenyltrifluoroborate
214.04
White
C7H7BF3KO
MFCD04112717
Authentic
≥96.0%

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning