missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium (4-Methoxyphenyl)trifluoroborate, TCI America™

Manufacturer:  TCI America P18205G

 View more versions of this product

Catalog No. P18205G



Specifications

Specifications

Potassium (4-Methoxyphenyl)trifluoroborate
≥98.0% (HPLC,T)
MFCD04112717
[B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+]
214.036
214.04
White
192863-36-8
C7H7BF3KO
XNYMCUFKZHRYID-UHFFFAOYSA-N
potassium;trifluoro-(4-methoxyphenyl)boranuide
23683762
Crystal-Powder at 20°C
5g

Chemical Identifiers

192863-36-8
214.036
XNYMCUFKZHRYID-UHFFFAOYSA-N
potassium;trifluoro-(4-methoxyphenyl)boranuide
C7H7BF3KO
MFCD04112717
23683762
[B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+]
Safety and Handling

Safety and Handling

TSCA : No

Documents