Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-(hydroxymethyl)phenyltrifluoroborate, 95%

$64.04 - $245.61

Chemical Identifiers

Chemical Name or Material Potassium 4-(hydroxymethyl)phenyltrifluoroborate
CAS 1015082-78-6
Molecular Formula C7H7BF3KO
Formula Weight 214.04
MDL Number MFCD08276800
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5188306

  SDS
alfa aesar™
H5188306
5g Each for $245.61

AAH5188303

  SDS
alfa aesar™
H5188303
1g Each for $64.04
Description & Specifications

Specifications

Formula Weight 214.04
Chemical Name or Material Potassium 4-(hydroxymethyl)phenyltrifluoroborate
Assay 95%
Molecular Formula C7H7BF3KO
MDL Number MFCD08276800
CAS 1015082-78-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 5g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.