Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-(furfurylaminocarbonyl)phenyltrifluoroborate

Chemical Identifiers

Chemical Name or Material Potassium 4-(furfurylaminocarbonyl)phenyltrifluoroborate
Molecular Formula C12H10BF3KNO2
Formula Weight 307.12
MDL Number MFCD11977716
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5114703

  SDS
alfa aesar™
H5114703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5114706

  SDS
alfa aesar™
H5114706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 307.12
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Quantity 1g
Chemical Name or Material Potassium 4-(furfurylaminocarbonyl)phenyltrifluoroborate
MDL Number MFCD11977716
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Molecular Formula C12H10BF3KNO2
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.