Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-formylphenyltrifluoroborate, 97%

$79.58 - $243.20

Chemical Identifiers

Chemical Name or Material Potassium 4-formylphenyltrifluoroborate
CAS 374564-36-0
Molecular Formula C7H5BF3KO
Formula Weight 212.02
MDL Number MFCD02093980
Synonym 4-Formylbenzenetrifluoroboric acid potassium salt; 4-Formylphenyltrifluoroboric acid potassium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1796903

  SDS
alfa aesar™
L1796903
1g Each for $79.58

AAL1796906

  SDS
alfa aesar™
L1796906
5g Each for $243.20
Description & Specifications

Specifications

Formula Weight 212.02
Chemical Name or Material Potassium 4-formylphenyltrifluoroborate
Assay 97%
MDL Number MFCD02093980
CAS 374564-36-0
Synonym 4-Formylbenzenetrifluoroboric acid potassium salt; 4-Formylphenyltrifluoroboric acid potassium salt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Quantity 1g
Molecular Formula C7H5BF3KO
Melting Point >300°C

Applications
It is employed in the synthesis of unsaturated organotrifluoroborates via Wittig and Horner-Wadsworth-Emmons Olefination.

Solubility
Soluble in water.

Notes
Air sensitive. Store away from air and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.