missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium 4-fluorophenyltrifluoroborate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1765503

 View more versions of this product

Catalog No. AAL1765503



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Potassium 4-fluorophenyltrifluoroborate
290°C to 292°C
MFCD01318170
7783544
PXKNUQDMYBUYSZ-UHFFFAOYSA-N
potassium;trifluoro-(4-fluorophenyl)boranuide
23677644
95%
192863-35-7
C6H4BF4K
1 g
potassium 4-fluorophenyltrifluoroborate, potassium trifluoro 4-fluorophenyl borate, potassium trifluoro 4-fluorophenyl boranuide, potassium 4-fluorophenyl trifluoroborate, 4-fluorophenyl-trifluoroborate potassium salt, 4-fluorophenyltrifluoroboric acid potassium salt, 4-fluorobenzenetrifluoroboric acid potassium salt, pubchem11422, potassiotrifluoro 4-fluorophenyl boron v, potassium trifluoro-4-fluorophenyl boranuide
[B-](C1=CC=C(C=C1)F)(F)(F)F.[K+]
202
202

Chemical Identifiers

192863-35-7
202
PXKNUQDMYBUYSZ-UHFFFAOYSA-N
23677644
[B-](C1=CC=C(C=C1)F)(F)(F)F.[K+]
C6H4BF4K
MFCD01318170
potassium 4-fluorophenyltrifluoroborate, potassium trifluoro 4-fluorophenyl borate, potassium trifluoro 4-fluorophenyl boranuide, potassium 4-fluorophenyl trifluoroborate, 4-fluorophenyl-trifluoroborate potassium salt, 4-fluorophenyltrifluoroboric acid potassium salt, 4-fluorobenzenetrifluoroboric acid potassium salt, pubchem11422, potassiotrifluoro 4-fluorophenyl boron v, potassium trifluoro-4-fluorophenyl boranuide
potassium;trifluoro-(4-fluorophenyl)boranuide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only