missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium 4-cyanophenyltrifluoroborate, 96%

Manufacturer:  Alfa Aesar™ H5318706

 View more versions of this product

Catalog No. AAH5318706



Specifications

Specifications

Potassium 4-cyanophenyltrifluoroborate
5g
C7H4BF3KN
potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanopheny trifluoroborate, potassium 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanophenyltrifluoroborat, trifluoro 4-cyanophenyl potassioboron v, potassium 4-cyanophenyl-trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium ion 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoro borate 1-
[B-](C1=CC=C(C=C1)C#N)(F)(F)F.[K+]
209.02
209.02
>300°C
850623-36-8
MFCD04115736
JEQAKQWNUUSKHI-UHFFFAOYSA-N
potassium;(4-cyanophenyl)-trifluoroboranuide
23716872
96%

Chemical Identifiers

850623-36-8
209.02
JEQAKQWNUUSKHI-UHFFFAOYSA-N
23716872
[B-](C1=CC=C(C=C1)C#N)(F)(F)F.[K+]
C7H4BF3KN
MFCD04115736
potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanopheny trifluoroborate, potassium 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanophenyltrifluoroborat, trifluoro 4-cyanophenyl potassioboron v, potassium 4-cyanophenyl-trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium ion 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoro borate 1-
potassium;(4-cyanophenyl)-trifluoroboranuide
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents