missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium (4-Cyanophenyl)trifluoroborate, TCI America™

Manufacturer:  TCI America P18151G

 View more versions of this product

Catalog No. P18151G



Specifications

Specifications

Potassium (4-Cyanophenyl)trifluoroborate
1g
C7H4BF3KN
potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanopheny trifluoroborate, potassium 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanophenyltrifluoroborat, trifluoro 4-cyanophenyl potassioboron v, potassium 4-cyanophenyl-trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium ion 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoro borate 1-
[B-](C1=CC=C(C=C1)C#N)(F)(F)F.[K+]
209.02
209.02
≥95.0% (N)
White
850623-36-8
MFCD04115736
JEQAKQWNUUSKHI-UHFFFAOYSA-N
potassium;(4-cyanophenyl)-trifluoroboranuide
23716872
Crystal-Powder at 20°C

Chemical Identifiers

850623-36-8
209.02
JEQAKQWNUUSKHI-UHFFFAOYSA-N
23716872
[B-](C1=CC=C(C=C1)C#N)(F)(F)F.[K+]
C7H4BF3KN
MFCD04115736
potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanopheny trifluoroborate, potassium 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium 4-cyanophenyltrifluoroborat, trifluoro 4-cyanophenyl potassioboron v, potassium 4-cyanophenyl-trifluoroboranuide, potassium 4-cyanophenyl trifluoroborate, potassium ion 4-cyanophenyl trifluoroboranuide, potassium 4-cyanophenyl trifluoro borate 1-
potassium;(4-cyanophenyl)-trifluoroboranuide
Safety and Handling

Safety and Handling

TSCA : No

Documents